Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108213.1 | NP_001101683.1 | 1 - 2 | 69 | 78 | |
XM_006244819.2 | XP_006244881.1 | 2 - 3 | 801 | 78 | ||
XM_006244820.2 | XP_006244882.1 | 2 - 3 | 1421 | 78 | ||
XM_006244821.2 | XP_006244883.1 | 1 - 2 | 603 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |