Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019151.1 | NP_062024.1 | 2 - 3 | 749 | 103 | |
GenBank mRNA | AF019624.1 | - | 2 - 3 | 749 | 103 | |
FQ224469.1 | - | 873 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |