Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108791.1 | NP_001102261.1 | 2 - 3 | 596 | 134 | |
XM_006244673.2 | XP_006244735.1 | 1 - 2 | 282 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |