Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnlz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130990.1 | NP_001124462.1 | 1 - 2 | 308 | 78 | |
GenBank mRNA | BQ205125.1 | - | 2 - 3 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |