Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004080.1 | NP_001004080.1 | 9 - 10 | 1350 | 73 | |
XM_006234039.2 | XP_006234101.1 | 9 - 10 | 1248 | 73 | ||
XM_006234040.1 | XP_006234102.1 | 10 - 11 | 1368 | 73 | ||
GenBank mRNA | BC079472.1 | - | 9 - 10 | 1350 | 7122783 | 73 |
FQ223877.1 | - | 10 - 11 | 1373 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |