Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114330.1 | NP_001107802.1 | 6 - 7 | 1978 | 64 | |
NM_017011.1 | NP_058707.1 | 6 - 7 | 1978 | 64 | ||
GenBank mRNA | M61099.1 | - | 6 - 7 | 1978 | 64 | |
X57569.1 | - | 6 - 7 | 1960 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Rattus norvegicus Probe spans exons |