Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bod1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770238.1 | XP_008768460.1 | 9 - 10 | 1931 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |