Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspbap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134419.2 | NP_599246.2 | 1 - 2 | 130 | 145 | |
GenBank mRNA | AF168362.1 | - | 1 - 2 | 119 | 145 | |
BC091125.1 | - | 1 - 2 | 130 | 7366096 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |