Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012145.1 | NP_001012145.1 | 11 - 12 | 1020 | 77 | |
GenBank mRNA | BC078948.1 | - | 11 - 12 | 1020 | 7109741 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |