Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127550.1 | NP_001121022.1 | 6 - 7 | 659 | 102 | |
GenBank mRNA | BC166791.1 | - | 6 - 7 | 659 | 7937077 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |