Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024899.1 | NP_001020070.1 | 10 - 11 | 1464 | 147 | |
XM_006231546.2 | XP_006231608.1 | 10 - 11 | 1515 | 147 | ||
XM_008760443.1 | XP_008758665.1 | 9 - 10 | 1524 | 147 | ||
GenBank mRNA | BC097447.1 | - | 10 - 11 | 1464 | 7456556 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |