Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108783.1 | NP_001102253.1 | 3 - 4 | 300 | 81 | |
XM_006243970.2 | XP_006244032.1 | 4 - 5 | 924 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |