Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031661.1 | NP_001026831.1 | 4 - 5 | 771 | 84 | |
XM_008771500.1 | XP_008769722.1 | 4 - 5 | 2528 | 84 | ||
XM_008771501.1 | XP_008769723.1 | 5 - 6 | 971 | 84 | ||
GenBank mRNA | BC099103.1 | - | 4 - 5 | 771 | 7460148 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |