Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013030.1 | NP_037162.1 | 5 - 6 | 587 | 84 | |
GenBank mRNA | BC078876.1 | - | 5 - 6 | 581 | 7109613 | 84 |
L13257.1 | - | 5 - 6 | 587 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |