Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017121.1 | NP_058817.1 | 6 - 7 | 864 | 76 | |
XM_008771439.1 | XP_008769661.1 | 5 - 6 | 779 | 76 | ||
GenBank mRNA | AF310623.2 | - | 6 - 7 | 864 | 76 | |
BC097263.1 | - | 6 - 7 | 847 | 7460052 | 76 | |
L14776.1 | - | 3 - 4 | 490 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |