Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024279.1 | NP_001019450.1 | 342 | 106 | ||
GenBank mRNA | AY963291.1 | - | 342 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Inventoried Probe spans exons |