Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115013.1 | NP_001108485.1 | 2 - 3 | 220 | 67 | |
GenBank mRNA | BI282694.1 | - | 3 - 4 | 67 | ||
CB607044.1 | - | 2 - 3 | 192 | 67 | ||
FQ223498.1 | - | 2 - 3 | 175 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |