Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749031.3 | XP_003749079.1 | 2 - 3 | 978 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |