Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033870.1 | NP_001029042.1 | 3 - 4 | 689 | 96 | |
NM_023102.2 | NP_075590.2 | 3 - 4 | 689 | 96 | ||
XM_006241000.2 | XP_006241062.1 | 3 - 4 | 1986 | 96 | ||
XM_006241001.2 | XP_006241063.1 | 3 - 4 | 694 | 96 | ||
XM_006241002.2 | XP_006241064.1 | 3 - 4 | 1984 | 96 | ||
XM_008765130.1 | XP_008763352.1 | 3 - 4 | 694 | 96 | ||
GenBank mRNA | BC072533.1 | - | 3 - 4 | 689 | 7098992 | 96 |
U22297.1 | - | 2 - 3 | 423 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |