Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133308.2 | NP_579842.2 | 2 - 3 | 1095 | 64 | |
GenBank mRNA | AF440691.2 | - | 2 - 3 | 1095 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |