Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108365.1 | NP_001101835.1 | 5 - 6 | 1208 | 126 | |
XM_006251340.2 | XP_006251402.1 | 4 - 5 | 1243 | 126 | ||
XM_008770332.1 | XP_008768554.1 | 3 - 4 | 1569 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |