Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105911.1 | NP_001099381.1 | 2 - 3 | 622 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |