Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230999.2 | XP_006231061.1 | 10 - 11 | 1025 | 70 | |
XM_006231000.2 | XP_006231062.1 | 10 - 11 | 893 | 70 | ||
GenBank mRNA | BC158834.1 | - | 10 - 11 | 987 | 7383724 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |