Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031008.2 | NP_112270.2 | 19 - 20 | 2619 | 142 | |
XM_008760010.1 | XP_008758232.1 | 19 - 20 | 2674 | 142 | ||
GenBank mRNA | BC081786.1 | - | 19 - 20 | 2619 | 7126468 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |