Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107510.1 | NP_001100980.1 | 4 - 5 | 434 | 76 | |
XM_006229051.2 | XP_006229113.1 | 6 - 7 | 782 | 76 | ||
XM_006229052.2 | XP_006229114.1 | 7 - 8 | 703 | 76 | ||
XM_006229053.2 | XP_006229115.1 | 6 - 7 | 818 | 76 | ||
XM_006229054.2 | XP_006229116.1 | 6 - 7 | 854 | 76 | ||
XM_008759366.1 | XP_008757588.1 | 5 - 6 | 595 | 76 | ||
XM_008759367.1 | XP_008757589.1 | 6 - 7 | 818 | 76 | ||
XM_008759368.1 | XP_008757590.1 | 6 - 7 | 815 | 76 | ||
XM_008759369.1 | XP_008757591.1 | 5 - 6 | 1647 | 76 | ||
GenBank mRNA | BC168932.1 | - | 6 - 7 | 859 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |