Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012687.1 | NP_036819.1 | 9 - 10 | 1259 | 92 | |
XM_008762759.1 | XP_008760981.1 | 9 - 10 | 1320 | 92 | ||
XM_008762760.1 | XP_008760982.1 | 7 - 8 | 2717 | 92 | ||
GenBank mRNA | D28773.1 | - | 9 - 10 | 1259 | 92 | |
D31798.1 | - | 9 - 10 | 1181 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |