Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139060.3 | NP_620691.2 | 5 - 6 | 1051 | 102 | |
XM_008768511.1 | XP_008766733.1 | 5 - 6 | 1056 | 102 | ||
GenBank mRNA | AB063114.1 | - | 5 - 6 | 1043 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |