Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021848.1 | NP_068620.1 | 9 - 10 | 1055 | 145 | |
GenBank mRNA | AF105368.1 | - | 9 - 10 | 1055 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |