Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100700.1 | NP_001094170.1 | 1 - 2 | 715 | 126 | |
GenBank mRNA | BC161900.1 | - | 1 - 2 | 715 | 8367121 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |