Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277177.1 | NP_001264106.1 | 3 - 4 | 775 | 143 | |
NM_001277178.1 | NP_001264107.1 | 3 - 4 | 775 | 143 | ||
GenBank mRNA | BC099835.1 | - | 3 - 4 | 518 | 143 | |
BC169044.1 | - | 3 - 4 | 775 | 9034472 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |