Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108309.1 | NP_001101779.1 | 10 - 11 | 1462 | 71 | |
GenBank mRNA | FQ212255.1 | - | 10 - 11 | 1372 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |