Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024488.1 | NP_077814.1 | 9 - 10 | 990 | 85 | |
GenBank mRNA | AF177476.1 | - | 9 - 10 | 990 | 85 | |
BC081793.1 | - | 8 - 9 | 912 | 7190258 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |