Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009622.1 | NP_001009622.1 | 2 - 3 | 140 | 135 | |
GenBank mRNA | BC088842.1 | - | 2 - 3 | 140 | 7303357 | 135 |
FQ217751.1 | - | 2 - 3 | 201 | 135 | ||
FQ219381.1 | - | 2 - 3 | 201 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |