Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017090.2 | NP_058786.2 | 7 - 8 | 1844 | 78 | |
XM_006232518.2 | XP_006232580.1 | 8 - 9 | 2436 | 78 | ||
XM_008761107.1 | XP_008759329.1 | 6 - 7 | 2467 | 78 | ||
XM_008761108.1 | XP_008759330.1 | 7 - 8 | 2210 | 78 | ||
GenBank mRNA | AB096020.1 | - | 6 - 7 | 1707 | 78 | |
BC085746.1 | - | 7 - 8 | 1844 | 7190419 | 78 | |
M57405.1 | - | 7 - 8 | 2163 | 78 | ||
U60835.1 | - | 7 - 8 | 2163 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |