Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025049.1 | NP_001020220.1 | 2 - 3 | 350 | 85 | |
XM_006237379.2 | XP_006237441.1 | 2 - 3 | 890 | 85 | ||
GenBank mRNA | BC079212.1 | - | 2 - 3 | 350 | 7131828 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |