Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763302.1 | XP_008761524.1 | 6 - 7 | 619 | 82 | |
XM_008763303.1 | XP_008761525.1 | 5 - 6 | 521 | 82 | ||
XM_008763304.1 | XP_008761526.1 | 5 - 6 | 636 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |