Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133591.2 | NP_598275.1 | 7 - 8 | 845 | 149 | |
XM_008767941.1 | XP_008766163.1 | 7 - 8 | 1206 | 149 | ||
XM_008767942.1 | XP_008766164.1 | 7 - 8 | 1271 | 149 | ||
XM_008767943.1 | XP_008766165.1 | 6 - 7 | 1119 | 149 | ||
GenBank mRNA | AF022774.1 | - | 6 - 7 | 771 | 149 | |
BC091126.1 | - | 7 - 8 | 845 | 7380477 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |