Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153629.1 | NP_705893.1 | 4 - 5 | 653 | 88 | |
XM_006246232.1 | XP_006246294.1 | 4 - 5 | 697 | 88 | ||
GenBank mRNA | AF077354.1 | - | 4 - 5 | 653 | 88 | |
FQ222433.1 | - | 5 - 6 | 861 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |