Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106717.1 | NP_001100187.1 | 4 - 5 | 773 | 102 | |
XM_006239896.1 | XP_006239958.1 | 3 - 4 | 600 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |