Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135899.1 | NP_001129371.1 | 3 - 4 | 546 | 122 | |
GenBank mRNA | BC127545.1 | - | 3 - 4 | 546 | 7321962 | 122 |
FQ211046.1 | - | 3 - 4 | 602 | 122 | ||
FQ219104.1 | - | 3 - 4 | 590 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |