Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053862.2 | NP_446314.2 | 4 - 5 | 479 | 109 | |
XM_006254166.2 | XP_006254228.1 | 4 - 5 | 745 | 109 | ||
XM_006254167.2 | XP_006254229.1 | 4 - 5 | 491 | 109 | ||
XM_006254168.1 | XP_006254230.1 | 4 - 5 | 518 | 109 | ||
GenBank mRNA | BC072488.1 | - | 4 - 5 | 479 | 7104910 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |