Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012891.2 | NP_037023.1 | 15 - 16 | 1553 | 75 | |
GenBank mRNA | BC087036.1 | - | 15 - 16 | 1574 | 7193535 | 75 |
D30647.1 | - | 15 - 16 | 1555 | 75 | ||
FQ210754.1 | - | 13 - 14 | 1756 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |