Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012891.2 | NP_037023.1 | 17 - 18 | 1695 | 58 | |
GenBank mRNA | BC087036.1 | - | 17 - 18 | 1716 | 7193535 | 58 |
D30647.1 | - | 17 - 18 | 1697 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |