Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mif4gd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014122.1 | NP_001014144.1 | 4 - 5 | 478 | 101 | |
XM_006247722.1 | XP_006247784.1 | 4 - 5 | 572 | 101 | ||
GenBank mRNA | BC079310.1 | - | 4 - 5 | 478 | 7134761 | 101 |
FQ227422.1 | - | 4 - 5 | 572 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |