Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107073.1 | NP_001100543.1 | 5 - 6 | 509 | 56 | |
XM_006247889.2 | XP_006247951.1 | 5 - 6 | 558 | 56 | ||
XM_008768477.1 | XP_008766699.1 | 5 - 6 | 558 | 56 | ||
GenBank mRNA | BC167031.1 | - | 5 - 6 | 491 | 7099085 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |