Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108278.1 | NP_001101748.1 | 3 - 4 | 355 | 95 | |
XM_006246743.1 | XP_006246805.1 | 4 - 5 | 476 | 95 | ||
XM_006246744.2 | XP_006246806.1 | 4 - 5 | 377 | 95 | ||
GenBank mRNA | FQ218736.1 | - | 4 - 5 | 336 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |