Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002730284.4 | XP_002730330.1 | 5 - 6 | 1587 | 77 | |
XM_006257246.2 | XP_006257308.1 | 4 - 5 | 1336 | 77 | ||
XM_006257247.1 | XP_006257309.1 | 6 - 7 | 662 | 77 | ||
XM_006257248.1 | XP_006257310.1 | 7 - 8 | 743 | 77 | ||
XM_006257250.2 | XP_006257312.1 | 6 - 7 | 617 | 77 | ||
XM_008773457.1 | XP_008771679.1 | 5 - 6 | 3996 | 77 | ||
XM_008773458.1 | XP_008771680.1 | 5 - 6 | 2728 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |