Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030020.1 | NP_001025191.1 | 5 - 6 | 585 | 106 | |
GenBank mRNA | BC099223.1 | - | 4 - 5 | 328 | 7388087 | 106 |
M12492.1 | - | 5 - 6 | 579 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |