Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009466.2 | NP_001009466.1 | 2 - 3 | 268 | 76 | |
XM_006239647.2 | XP_006239709.1 | 2 - 3 | 370 | 76 | ||
GenBank mRNA | AY053516.1 | - | 2 - 3 | 269 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |