Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172034.2 | NP_742031.1 | 7 - 8 | 735 | 65 | |
GenBank mRNA | BC087675.1 | - | 7 - 8 | 735 | 7319293 | 65 |
M69056.1 | - | 7 - 8 | 749 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |